missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Toluene, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8%, Honeywell

Supplier:  Honeywell Chemicals LP3474

 View more versions of this product

Catalog No. 6001724


Explore available promotions

Description

Description

Specifications

Specifications

Toluene
108-88-3
0.867 g/cm3
4 x 4 L
Glass Bottle
C7H8
UN1294
635760
5.2% in Water at 20°C
CC1=CC=CC=C1
92.14
CHEBI:17578
>99.5%
ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8%
100.0000%
110.6°C
4°C
by GC Analysis
MFCD00008512
0.03% max.
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
ACS Reagent

Chemical Identifiers

108-88-3
92.14
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CHEBI:17578
CC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene

SDS
Documents

Documents

Promotions

Promotions