Learn More
Toluene, anhydrous, 99.8%, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
$60.26 - $119.25
Chemical Identifiers
CAS | 108-88-3 |
---|---|
Molecular Formula | C7H8 |
Molecular Weight (g/mol) | 92.14 |
MDL Number | MFCD00008512 |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
Synonym | methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene |
PubChem CID | 1140 |
ChEBI | CHEBI:17578 |
IUPAC Name | toluene |
SMILES | CC1=CC=CC=C1 |
Description
Toluene is added to gasoline, used to produce benzene, and used as a solvent. It is used in aviation and automotive fuels, as a solvent, and to make other chemicals. Toluene has also been used as a coolant for its good heat transfer capabilities in sodium cold traps used in nuclear reactor system loops.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
108-88-3 | |
92.14 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
1140 | |
toluene |
C7H8 | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CHEBI:17578 | |
CC1=CC=CC=C1 |
Specifications
-93°C | |
Colorless | |
111°C | |
4°C (40°F) | |
ChemSeal™ Bottle | |
1.4967 | |
C6H5CH3 | |
UN1294 | |
14,9529 | |
Insoluble in water. | |
CC1=CC=CC=C1 | |
92.14 | |
CHEBI:17578 | |
99.8% |
108-88-3 | |
0.867 g/mL | |
100 mL | |
Petroleum-like | |
Liquid | |
C7H8 | |
MFCD00008512 | |
635760 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
1140 | |
92.14 | |
Toluene, Anhydrous, over molecular sieves |
Safety and Handling
GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336-H361d-H373
DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: TOLUENE
EINECSNumber : 203-625-9
RTECSNumber : XS5250000
TSCA : Yes
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only