missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Toluene (HPLC), Fisher Chemical™

Supplier:  Fisher Chemical BPT290SK4

 View more versions of this product

Catalog No. T290SK-4



Explore available promotions

Description

Description

Specifications

Specifications

Toluene
Also meets ACS specifications
1 AU max. at 286nm
0.866g/cm³
4 L
Safe-Cote™ Amber Glass 
Mobile phase for HPLC applications
10ppm max.
C7H8
29mbar at 20°C
0.02% max.
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
HPLC
-95°C
108-88-3
Colorless
111°C
Pass Test
Liquid
1.4930 to 1.4950 (at 25°C)
99.8% min by GC
MFCD00008512
0.6 mPa/s at 20°C
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CC1=CC=CC=C1
92.14
CHEBI:17578
≥99.8%

Chemical Identifiers

108-88-3
92.14
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CHEBI:17578
CC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

Signal Word: DANGER!

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Promotions

Promotions