Invitrogen™ TOP10 Electrocomp™ Kit DFS Item
TOP10 Electrocomp E. coli cells are provided at a transformation efficiency of 1 x 1010 cfu/μg supercoiled DNA and are ideal for high-efficiency cloning and plasmid propagation.
Supplier:  Invitrogen™ C66411
 View more versions of this product
Catalog No. C66411
Explore available promotions
Description
Description
TOP10 Electrocomp E. coli cells are provided at a transformation efficiency of 1 x 1010 cfu/μg supercoiled DNA and are ideal for high-efficiency cloning and plasmid propagation. This strain is used in wide range of applications and allows stable replication of high-copy number plasmids and maintenance of large plasmids. 
The TOP10 genotype is similar to the DH10B strain and features:
hsdR for efficient transformation of unmethylated DNA from PCR amplifications
mcrA for efficient transformation of methylated DNA from genomic preparations
lacZΔM15 for blue/white color screening of recombinant clones
endA1 for cleaner preparations of DNA and better results in downstream applications due to the elimination of non-specific digestion by Endonuclease I
recA1 for reduced occurrence of non-specific recombination in cloned DNA
Notes
Please see the product manual for reaction scale recommendations. A high-voltage electroporation apparatus is required.
Genotype
F- mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK rpsL endA1 nupG
Find the strain and format that fits your needs
• TOP10 is available in chemically competent and electrocompetent cell formats.
• Choose MultiShot competent cells for high-throughput cloning applications.
Specifications
Specifications
Electrocompetent Cells
Yes (endA1)
High Efficiency (>1 x 1010 cfu/μg)
Yes (Streptomycin)
Yes (lacZΔM15)
May be used for plasmids >20 kb
Yes (recA1)
No
Tube
10 x 100 μL
No
Yes (mcrA)
• 10 x 100 μL TOP10 Electrocomp E. coli
• 50 μL pUC19 vector (10 pg/μL)
Store at –80°C.
• 15 mL S.O.C. Medium
Store at room temperature.
Not suitable for cloning unstable DNA
Low
No
Dry Ice
E. coli (K12)
Electrocomp™
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.