missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell TouchNTuff™ 92-600 Disposable Chemical Resistant Nitrile Gloves
Soft nitrile disposable gloves provide high levels of comfort and splash resistance against hazardous chemicals.
Supplier: Ansell 9260011080
Description
Soft nitrile disposable gloves provide high levels of comfort and splash resistance against hazardous chemicals.
- Made from 100% nitrile
- Resistant to nuisance punctures, nicks, cuts and abrasions
- Comfortable thin mil construction
- Rolled cuffs for easy donning and doffing
- Chemical resistant
Specifications
ASTM D6319, FDA 21 CFR 177.2600, ASTM F1671 | |
No | |
Single-use | |
0.14 mm | |
Yes | |
4.7 mil | |
Medium | |
0.14 mm | |
Type B | |
Bacteria, Fungi WITH Virus Protection | |
Unlined | |
92 600 M 585835 | |
9.5 in. |
Category III, EN 420:2003 + A1:2009 | |
Green | |
5.5 mil | |
Ambidextrous | |
Nitrile | |
0.12 mm | |
5.5 mil | |
No | |
JKPT | |
Beaded/Rolled | |
ISO 9001 | |
TouchNTuff™ 92-600 | |
24 cm |