missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ansell TouchNTuff™ 92-600 Disposable Chemical Resistant Nitrile Gloves Edge
Edge

Soft nitrile disposable gloves provide high levels of comfort and splash resistance against hazardous chemicals.

Supplier:  Ansell 9260011090

 View more versions of this product

Catalog No. 18-999-2748C



Edge
Explore available promotions

Description

Description

Soft nitrile disposable gloves provide high levels of comfort and splash resistance against hazardous chemicals.

  • Made from 100% nitrile
  • Resistant to nuisance punctures, nicks, cuts and abrasions
  • Comfortable thin mil construction
  • Rolled cuffs for easy donning and doffing
  • Chemical resistant
Specifications

Specifications

ASTM D6319, FDA 21 CFR 177.2600, ASTM F1671
No
Single-use
0.14 mm
Yes
4.7 mil
Large
0.14 mm
Type B
Bacteria, Fungi WITH Virus Protection
Unlined
92 600 L 585836
9.5 in.
Category III, EN 420:2003 + A1:2009
Green
5.5 mil
Ambidextrous
Nitrile
0.12 mm
5.5 mil
No
JKPT
Beaded/Rolled
ISO 9001
TouchNTuff™ 92-600
24 cm
SDS
Documents

Documents

Promotions

Promotions