missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TOX Monoclonal Antibody (TXRX10), eFluor™ 660, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Supplier:  Thermo Fisher Scientific 50650282

 View more versions of this product

Catalog No. 501124423


Explore available promotions

Description

Description

Description: This TXRX10 monoclonal antibody reacts with human and mouse TOX, which is also known as Thymocyte selection-associated HMG box protein. This DNA-binding nuclear factor is expressed in thymocytes, T lymphocytes, NK cells, and lymphoid tissue-inducer (LTi) cells. TOX expression in thymocytes is upregulated during beta-selection and positive selection. Studies using Tox knockout mice have demonstrated a critical role for TOX in CD4+ T cell development as a regulator of the CD4loCD8lo to CD4+CD8lo transition. Moreover, TOX has been shown to be involved in the development of NK and LTi cells. While its regulation remains unclear, TOX expression has been shown to be induced by calcineurin-mediated TCR signaling in thymocytes during positive selection. Applications Reported: This TXRX10 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This TXRX10 antibody has been tested by intracellular staining and flow cytometric analysis of mouse thymocytes using the Foxp3/Transcriptin Factor Buffer Set (cat 00-5523) and protocol. Refer to Best Protocols- (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.

TOX (thymocyte selection-associated high mobility group (HMG) box protein) is a member of the HMG box family of DNA-binding proteins and likely plays a role in the regulation of T-cell development. It is a 526 amino acid nuclear protein and the expression of TOX is upregulated by pre-T cell receptor (pre-TCR) and TCR activation in immature thymocytes. TOX-mediated positive selection is associated with up-regulation of RUNX3 and is calcineurin dependent. TOX-dependent transition to the CD4+CD8 stage is required for development of class II major histocompatibility complex-specific T cells.
Specifications

Specifications

TOX
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
O94900, Q66JW3
TOX
Affinity chromatography
RUO
252838, 9760
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
TXRX10
eFluor 660
TOX
1700007F02Rik; KIAA0808; thymocyte selection associated high mobility group box; thymocyte selection-associated high mobility group box; thymocyte selection-associated high mobility group box protein TOX; thymocyte selection-associated HMG box; thymus high mobility group box protein TOX; TOX; TOX1
Rat
100 μg
Primary
Human, Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only.