FUJIFILM Wako Toxinometer™ ET-7000/E, Part 11 Set
Computer-operated kinetic tube reader to perform all 3 LAL testing methodologies, contains analysis module, software, laptop computer and system validation documents
Supplier:  FUJIFILM Wako 29935821
Catalog No. 12-333-043
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Single assay module can simultaneously measure up to 16 samples
  • Expansion modules available (up to 8) to extend number of samples in multiples of 16
  • Single-test configuration avoids hot wells phenomenon associated with microplate readers by evenly distributing temperature in all single wells
  • Reduces chance of contamination due to single-well formats
  • Ideal for users converting from gel clot method to kinetic turbidimetric assays
  • 220 to 240 ± 10% VAC
Specifications
Specifications
Kinetic Tube Reader
Performing LAL testing methodologies, For users converting from gel clot method to kinetic turbidimetric assays
Analysis module, Software, Laptop computer, System validation documents