Eisco™ TPX Graduated Cylinders - Class A
Class A TPX graduated cylinder featuring raised/ molded graduations, octagonal base for maximum stability, and spout. Easy to read with amazing clarity.
$13.59 - $119.71
Specifications
Class Class A
Disposable Reusable
Quantity 1
Autoclavable Autoclavable
Base Type Octagonal
View More Specs
Products 7
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S88817
View Documents
Eisco™
CH0354P
10 mL
Each for $13.59
S88818
View Documents
Eisco™
CH0354Q
25 mL
Each for $17.15
S88819
View Documents
Eisco™
CH0354R
50 mL
Each for $20.18
S88820
View Documents
Eisco™
CH0354S
100 mL
Each for $33.09
S88821
View Documents
Eisco™
CH0354T
250 mL
Each for $47.62
S88822
View Documents
Eisco™
CH0354U
500 mL
Each for $119.71
S88823
View Documents
Eisco™
CH0354V
1000 mL
Each for $99.94
Description
Description
  • High quality TPX plastic graduated cylinders
  • Single piece construction
  • Superior optical clarity
  • Easy to read, molded graduations
  • Good chemical resistance
  • Octagonal base for maximum stability
  • Spout for easy pouring
  • Class A
  • Autoclavable
  • Warranty
    1 year limited manufacturer warranty
    Specifications
    Specifications
    Class A
    1
    Octagonal
    Reusable
    Autoclavable
    Graduated Measuring Cylinder
    Videos
    SDS
    missing translation for 'documents'
    missing translation for 'documents'