missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Traceable™ One-Shot™ Conductivity Calibration Standards Certified Reference Material Producer (CRM), Fisherbrand™
Ideal for lab or field applications
$93.83 - $260.65
Specifications
Chemical Name or Material | Conductivity Standard |
---|---|
Packaging | 6/Pk. |
Color | Clear |
For Use With (Equipment) | Conductivity Meter |
Solubility | Soluble in water |
Catalog Number | Mfr. No. | Quantity | Conductivity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Conductivity | Price | Quantity | |||||
06-664-25
|
N/A | 100 mL | 5 μ℧ |
Pack of 6 Each for $260.65
|
|
|||||
09-328-6
![]() |
N/A | 100 mL | 10 μ℧ |
Pack of 6 Each for $98.72
|
![]() |
|||||
09-328-7
![]() |
N/A | 6 x 100 mL | 100 μ℧ |
Pack of 6 Each for $93.83
|
![]() |
|||||
09-328-8
![]() |
N/A | 100 mL | 1000 μ℧ |
Pack of 6 Each for $93.83
|
![]() |
|||||
09-328-12
|
N/A | 6 x 100 mL | 1413 μ℧ |
Pack of 6 Each for $94.00
|
|
|||||
09-328-9
|
N/A | 100 mL | 10,000 μ℧ |
Pack of 6 Each for $94.00
|
|
|||||
09-328-10
|
N/A | 100 mL | 100,000 μ℧ |
Pack of 6 Each for $93.87
|
|
|||||
09-328-20
|
N/A | 6 x 100 mL | 10, 100, 1000, 1413, 10,000, 100,000 μ℧ |
Pack of 6 Each for $98.00
|
|
|||||
Description
Ideal for lab or field applications
Single-use conductivity standards calibrate all conductivity meters and probes for maximum accuracy. Eliminate concern about external container contamination. Calibration is made in the standard vial.
Calibration within container — zero contamination
- Traceable to NIST (National Institute of Standards and Technology)
- Accuracy: certificate stated uncertainty or ±0.25 microsiemen or ±0.25%
- 100% compatible with all conductivity meters
- A2LA accreditations ISO 34 and ISO 17025 plus ISO 9001
- Includes an individual temperature chart and step-by-step calibration instructions
- Units labeled (micromhos/microsiemens, ohms, parts per million/TDS)
- Single use Eliminates external contamination concerns Each standard contains 100mL
Assortment (09-328-20) contains one each of 09-328-6, -7, -8, -9, -10, and -12
Order Info
Each bottle contains 100mL
Specifications
Conductivity Standard | |
Clear | |
Soluble in water |
6/Pk. | |
Conductivity Meter |