Fisherbrand™ Traceable™ Platinum Ultra-Accurate Digital Thermometer
High-precision unit is designed for years of reliable service—even in the severest environments.
 View more versions of this product
Catalog No. 15-081-102
Explore available promotions
Includes: Probe, cable, Plastic carrying case, traceable certificate and 9V battery
Description
Description
High-precision unit is designed for years of reliable service—even in the severest environments. Fisher Scientific™ Traceable™ Platinum Ultra-Accurate Digital Thermometers have platinum sensor which ensures linear response and high accuracy across the entire range. Ideal for foods, pharmaceuticals, petroleum products, wastewater, gas systems, soil, ovens and electronics manufacturing.
  • Platinum sensor insures linear response and high accuracy across the entire range
  • Ideal for foods, pharmaceuticals, petroleum products, wastewater, gas systems, soil, ovens, electronics manufacturing
  • Membrane-covered keys are splash-proof and easy to operate
  • HOLD allows user to freeze display for reading later
  • Displays data in °F or °C
  • Computer output supplied with unit
  • Handle Probe: (range –200 to 500°C): Fast-response, waterproof, 4-wire platinum/stainless-steel 316 probe, 0.16-inch diameter x 7-inch stem length, overall length 11.5 inches; probe tip penetration of 0.3-inch, 6-foot cable
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Specifications
Specifications
ISO 17025, A2LA, NIST
1.27 cm
0.4 cm
Measuring temperatures in gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, foods and wastewater
13.97 cm
6 ft.
292 mm
17.7 cm
Handle Probe
8 oz.
3.5 in.
932°F
-328°F
-200°C to +500°C
Digital LCD
0.5 in.
0.16 in.
11.5 x 3.5 x 5.5 in. (29.21 x 8.89 x 13.97 cm)
5.5 in.
Stainless Steel probe with handle, cable, computer output capabilities (software package must be purchased separately), Plastic carrying case, traceable certificate and 9V battery
7 in.
182.88 cm
Platinum, Stainless Steel
0.001°C
226.7 g
11.5 in.
500°C
-200°C
±0.05°C
8.89 cm