missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-1,2-Dibromocyclohexane, 99%, Thermo Scientific™

$278.96 - $278.96

Chemical Identifiers

CAS 7429-37-0
Molecular Formula C6H10Br2
Molecular Weight (g/mol) 241.95
MDL Number MFCD00003820
InChI Key CZNHKZKWKJNOTE-PHDIDXHHSA-N
Synonym trans-1,2-dibromocyclohexane, 1r,2r-1,2-dibromocyclohexane, +/--trans-1,2-dibromocyclohexane, cyclohexane,1,2-dibromo-,trans, trans-1,2-dibromcyclohexan, 1beta,2alpha-dibromocyclohexane, cyclohexane,2-dibromo-, trans, 1r,2r-1,2-bis bromanyl cyclohexane
PubChem CID 98506
IUPAC Name (1R,2R)-1,2-dibromocyclohexane
SMILES Br[C@@H]1CCCC[C@H]1Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC112770250
SDS View Documents
Thermo Scientific Chemicals
112770250
25 mL Glass bottle
Each for $278.96
 
Description

Description

Specifications

Chemical Identifiers

7429-37-0
241.95
CZNHKZKWKJNOTE-PHDIDXHHSA-N
98506
Br[C@@H]1CCCC[C@H]1Br
C6H10Br2
MFCD00003820
trans-1,2-dibromocyclohexane, 1r,2r-1,2-dibromocyclohexane, +/--trans-1,2-dibromocyclohexane, cyclohexane,1,2-dibromo-,trans, trans-1,2-dibromcyclohexan, 1beta,2alpha-dibromocyclohexane, cyclohexane,2-dibromo-, trans, 1r,2r-1,2-bis bromanyl cyclohexane
(1R,2R)-1,2-dibromocyclohexane

Specifications

7429-37-0
1.7800g/mL
>110°C
99%
C6H10Br2
C6H10Br2
25 mL
1.78
CZNHKZKWKJNOTE-PHDIDXHHSA-N
(1R,2R)-1,2-dibromocyclohexane
98506
99%
trans-1, 2-Dibromocyclohexane, 99%
Yellow
145°C (100.0 mmHg)
Authentic
Glass bottle
1.5510 to 1.553
MFCD00003820
05, 24
trans-1,2-dibromocyclohexane, 1r,2r-1,2-dibromocyclohexane, +/--trans-1,2-dibromocyclohexane, cyclohexane,1,2-dibromo-,trans, trans-1,2-dibromcyclohexan, 1beta,2alpha-dibromocyclohexane, cyclohexane,2-dibromo-, trans, 1r,2r-1,2-bis bromanyl cyclohexane
Br[C@@H]1CCCC[C@H]1Br
241.95
241.95
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-070-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only