missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dichloroethylene, 99+%, stabilized

CAS: 156-60-5 | C2H2Cl2 | 96.94 g/mol

$180.71 - $180.71

Chemical Identifiers

CAS 156-60-5
Molecular Formula C2H2Cl2
Molecular Weight (g/mol) 96.94
MDL Number MFCD00062942
InChI Key KFUSEUYYWQURPO-OWOJBTEDSA-N
Synonym trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform
PubChem CID 638186
ChEBI CHEBI:29027
IUPAC Name (E)-1,2-dichloroethene
SMILES Cl\C=C\Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC406840250
SDS View Documents
Thermo Scientific Chemicals
406840250
25 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

156-60-5
96.94
KFUSEUYYWQURPO-OWOJBTEDSA-N
638186
(E)-1,2-dichloroethene
C2H2Cl2
MFCD00062942
trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform
CHEBI:29027
Cl\C=C\Cl

Specifications

156-60-5
Colorless
48°C
Authentic
Glass bottle
1.4450 to 1.447
MFCD00062942
01, 188
15, 87
Solubility in water: immiscible
Cl\C=C\Cl
96.94
638186
96.94
Liquid
-50°C
1.2600g/mL
6°C
99% min. (GC)
C2H2Cl2
CHClCHCl
25 g
1.26
trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform
KFUSEUYYWQURPO-OWOJBTEDSA-N
(E)-1,2-dichloroethene
85 to 115ppm MEHQ
CHEBI:29027
99+%
trans-1, 2-Dichloroethylene, 99.7%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Harmful if swallowed.

GHS P Statement
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away

GHS Signal Word: Danger

EINECSNumber : 205-860-2

RTECSNumber : KV9400000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only