missing translation for 'onlineSavingsMsg'
Learn More

trans-1,4-Cyclohexanedicarboxylic Acid 97.0+%, TCI America™

$89.48 - $758.67

Chemical Identifiers

CAS 619-82-9
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00066203
InChI Key PXGZQGDTEZPERC-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedicarboxylic acid, trans-1,4-cyclohexanedicarboxylic acid, cis-1,4-cyclohexanedicarboxylic acid, cis-cyclohexane-1,4-dicarboxylic acid, trans-cyclohexane-1,4-dicarboxylic acid, hexahydroterephthalic acid, trans-hexahydroterephthalic acid, trans-1,4-cyclohexanedicarboxybic acid, cis-hexahydroterephthalic acid, 1,4-dicarboxycyclohexane
PubChem CID 14106
IUPAC Name cyclohexane-1,4-dicarboxylic acid
SMILES C1CC(CCC1C(=O)O)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C047525G
View Documents
TCI America
C047525G
25 g N/A
Only null left
 
C0475100G
View Documents
TCI America
C0475100G
100 g N/A
Only null left
 
C0475500G
View Documents
TCI America
C0475500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

619-82-9
172.18
PXGZQGDTEZPERC-UHFFFAOYSA-N
14106
C1CC(CCC1C(=O)O)C(=O)O
C8H12O4
MFCD00066203
1,4-cyclohexanedicarboxylic acid, trans-1,4-cyclohexanedicarboxylic acid, cis-1,4-cyclohexanedicarboxylic acid, cis-cyclohexane-1,4-dicarboxylic acid, trans-cyclohexane-1,4-dicarboxylic acid, hexahydroterephthalic acid, trans-hexahydroterephthalic acid, trans-1,4-cyclohexanedicarboxybic acid, cis-hexahydroterephthalic acid, 1,4-dicarboxycyclohexane
cyclohexane-1,4-dicarboxylic acid

Specifications

619-82-9
C8H12O4
25 g
PXGZQGDTEZPERC-UHFFFAOYSA-N
cyclohexane-1,4-dicarboxylic acid
14106
≥97.0% (GC,T)
trans-1,4-Cyclohexanedicarboxylic Acid
White
MFCD00066203
1,4-cyclohexanedicarboxylic acid, trans-1,4-cyclohexanedicarboxylic acid, cis-1,4-cyclohexanedicarboxylic acid, cis-cyclohexane-1,4-dicarboxylic acid, trans-cyclohexane-1,4-dicarboxylic acid, hexahydroterephthalic acid, trans-hexahydroterephthalic acid, trans-1,4-cyclohexanedicarboxybic acid, cis-hexahydroterephthalic acid, 1,4-dicarboxycyclohexane
C1CC(CCC1C(=O)O)C(=O)O
172.18
172.18
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2423&(3)-2446

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'