missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-1,4-Cyclohexanedimethanol 98.0+%, TCI America™
$97.89 - $293.65
Chemical Identifiers
CAS | 3236-48-4 |
---|---|
Molecular Formula | C8H16O2 |
Molecular Weight (g/mol) | 144.21 |
MDL Number | MFCD00001512,MFCD00066360 |
InChI Key | YIMQCDZDWXUDCA-UHFFFAOYSA-N |
Synonym | 1,4-cyclohexanedimethanol, trans-1,4-cyclohexanedimethanol, chdm, 1,4-bis hydroxymethyl cyclohexane, rikabinol dm, cyclohexane-1,4-dimethanol, 1,4-chidm, 1,4-dimethylolcyclohexane, 4-hydroxymethyl cyclohexyl methanol, 1,4-cyclohexanedimethanol, cis |
PubChem CID | 7735 |
IUPAC Name | [4-(hydroxymethyl)cyclohexyl]methanol |
SMILES | OCC1CCC(CO)CC1 |
Chemical Identifiers
3236-48-4 | |
144.21 | |
YIMQCDZDWXUDCA-UHFFFAOYSA-N | |
7735 | |
OCC1CCC(CO)CC1 |
C8H16O2 | |
MFCD00001512,MFCD00066360 | |
1,4-cyclohexanedimethanol, trans-1,4-cyclohexanedimethanol, chdm, 1,4-bis hydroxymethyl cyclohexane, rikabinol dm, cyclohexane-1,4-dimethanol, 1,4-chidm, 1,4-dimethylolcyclohexane, 4-hydroxymethyl cyclohexyl methanol, 1,4-cyclohexanedimethanol, cis | |
[4-(hydroxymethyl)cyclohexyl]methanol |
Specifications
3236-48-4 | |
White | |
C8H16O2 | |
5 g | |
YIMQCDZDWXUDCA-UHFFFAOYSA-N | |
[4-(hydroxymethyl)cyclohexyl]methanol | |
7735 | |
≥98.0% (GC) | |
trans-1,4-Cyclohexanedimethanol |
67°C | |
164°C | |
MFCD00001512,MFCD00066360 | |
1,4-cyclohexanedimethanol, trans-1,4-cyclohexanedimethanol, chdm, 1,4-bis hydroxymethyl cyclohexane, rikabinol dm, cyclohexane-1,4-dimethanol, 1,4-chidm, 1,4-dimethylolcyclohexane, 4-hydroxymethyl cyclohexyl methanol, 1,4-cyclohexanedimethanol, cis | |
OCC1CCC(CO)CC1 | |
144.21 | |
144.21 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2326
TSCA : Yes