missing translation for 'onlineSavingsMsg'
Learn More

trans-1,4-Dimethylcyclohexane 95.0+%, TCI America™

$180.81 - $479.06

Chemical Identifiers

CAS 2207-04-7
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00064955
InChI Key QRMPKOFEUHIBNM-UHFFFAOYSA-N
Synonym trans-Hexahydro-p-xylene
PubChem CID 11523
IUPAC Name 1,4-dimethylcyclohexane
SMILES CC1CCC(CC1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D02715ML
View Documents
TCI America
D02715ML
5 mL
Each for $180.81
Only null left
 
D027125ML
View Documents
TCI America
D027125ML
25 mL
Each for $479.06
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2207-04-7
112.22
QRMPKOFEUHIBNM-UHFFFAOYSA-N
11523
CC1CCC(CC1)C
C8H16
MFCD00064955
trans-Hexahydro-p-xylene
1,4-dimethylcyclohexane

Specifications

2207-04-7
119°C
MFCD00064955
2263
QRMPKOFEUHIBNM-UHFFFAOYSA-N
1,4-dimethylcyclohexane
11523
≥95.0% (GC)
trans-1,4-Dimethylcyclohexane
Colorless
C8H16
5 mL
trans-Hexahydro-p-xylene
CC1CCC(CC1)C
112.22
112.22
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2227

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'