missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane, 97%, Thermo Scientific Chemicals
$451.07 - $1274.67
Chemical Identifiers
CAS | 1212405-03-2 |
---|---|
Molecular Formula | C12H22BrNO2 |
Molecular Weight (g/mol) | 292.217 |
MDL Number | MFCD12022626 |
InChI Key | XJDUGNDNCQMMBK-NXEZZACHSA-N |
Synonym | tert-butyl trans-3-bromomethyl-cyclohexylcarbamate, t-butyl trans-3-bromomethyl cyclohexyl carbamate, tert-butyl n-1r,3r-3-bromomethyl cyclohexyl carbamate |
PubChem CID | 46172881 |
IUPAC Name | tert-butyl N-[(1R,3R)-3-(bromomethyl)cyclohexyl]carbamate |
SMILES | CC(C)(C)OC(=O)NC1CCCC(C1)CBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH62670MD
|
Thermo Scientific Chemicals
H62670MD |
250 mg |
Each for $451.07
|
|
|||||
AAH6267003
|
Thermo Scientific Chemicals
H6267003 |
1 g |
Each for $1,274.67
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
NotesStable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
1212405-03-2 | |
292.217 | |
XJDUGNDNCQMMBK-NXEZZACHSA-N | |
46172881 | |
CC(C)(C)OC(=O)NC1CCCC(C1)CBr |
C12H22BrNO2 | |
MFCD12022626 | |
tert-butyl trans-3-bromomethyl-cyclohexylcarbamate, t-butyl trans-3-bromomethyl cyclohexyl carbamate, tert-butyl n-1r,3r-3-bromomethyl cyclohexyl carbamate | |
tert-butyl N-[(1R,3R)-3-(bromomethyl)cyclohexyl]carbamate |
Specifications
1212405-03-2 | |
MFCD12022626 | |
XJDUGNDNCQMMBK-NXEZZACHSA-N | |
tert-butyl N-[(1R,3R)-3-(bromomethyl)cyclohexyl]carbamate | |
292.217 | |
292.21 | |
trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane |
C12H22BrNO2 | |
tert-butyl trans-3-bromomethyl-cyclohexylcarbamate, t-butyl trans-3-bromomethyl cyclohexyl carbamate, tert-butyl n-1r,3r-3-bromomethyl cyclohexyl carbamate | |
CC(C)(C)OC(=O)NC1CCCC(C1)CBr | |
250 mg | |
46172881 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only