missing translation for 'onlineSavingsMsg'
Learn More

trans-1-(Boc-amino)-4-ethynylcyclohexane, 97%

CAS: 947141-86-8 | C13H21NO2 | 223.32 g/mol

$250.67 - $707.89

Chemical Identifiers

CAS 947141-86-8
Molecular Formula C13H21NO2
Molecular Weight (g/mol) 223.32
MDL Number MFCD12963985
InChI Key NZTBBJYAWNVCOE-UHFFFAOYSA-N
Synonym tert-butyl trans-4-ethynylcyclohexylcarbamate, trans-1-boc-amino-4-ethynylcyclohexane, tert-butyl n-4-ethynylcyclohexyl carbamate, nztbbjyawnvcoe-xypyzodxsa-n, tert-butyl trans 4-ethynylcyclohexylcarbamate, tert-butyl trans-4-ethynylcyclohexanecarbamate, tert-butyl trans-4-ethynylcyclohexyl carbamate, trans-4-ethynyl-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl n-1r,4r-4-ethynylcyclohexyl carbamate, 1alpha,4beta-4-ethynylcyclohexyl carbamic acid tert-butyl ester
PubChem CID 57538360
IUPAC Name tert-butyl N-(4-ethynylcyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62905MD
View Documents
Thermo Scientific Chemicals
H62905MD
250 mg
Each for $250.67
Only null left
 
AAH6290503
View Documents
Thermo Scientific Chemicals
H6290503
1 g
Each for $707.89
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

947141-86-8
223.32
NZTBBJYAWNVCOE-UHFFFAOYSA-N
57538360
CC(C)(C)OC(=O)NC1CCC(CC1)C#C
C13H21NO2
MFCD12963985
tert-butyl trans-4-ethynylcyclohexylcarbamate, trans-1-boc-amino-4-ethynylcyclohexane, tert-butyl n-4-ethynylcyclohexyl carbamate, nztbbjyawnvcoe-xypyzodxsa-n, tert-butyl trans 4-ethynylcyclohexylcarbamate, tert-butyl trans-4-ethynylcyclohexanecarbamate, tert-butyl trans-4-ethynylcyclohexyl carbamate, trans-4-ethynyl-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl n-1r,4r-4-ethynylcyclohexyl carbamate, 1alpha,4beta-4-ethynylcyclohexyl carbamic acid tert-butyl ester
tert-butyl N-(4-ethynylcyclohexyl)carbamate

Specifications

947141-86-8
MFCD12963985
NZTBBJYAWNVCOE-UHFFFAOYSA-N
tert-butyl N-(4-ethynylcyclohexyl)carbamate
223.32
223.31
trans-1-(Boc-amino)-4-ethynylcyclohexane
C13H21NO2
tert-butyl trans-4-ethynylcyclohexylcarbamate, trans-1-boc-amino-4-ethynylcyclohexane, tert-butyl n-4-ethynylcyclohexyl carbamate, nztbbjyawnvcoe-xypyzodxsa-n, tert-butyl trans 4-ethynylcyclohexylcarbamate, tert-butyl trans-4-ethynylcyclohexanecarbamate, tert-butyl trans-4-ethynylcyclohexyl carbamate, trans-4-ethynyl-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl n-1r,4r-4-ethynylcyclohexyl carbamate, 1alpha,4beta-4-ethynylcyclohexyl carbamic acid tert-butyl ester
CC(C)(C)OC(=O)NC1CCC(CC1)C#C
250 mg
57538360
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only