missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-2,4-Dichlorocinnamic Acid 98.0+%, TCI America™
$98.75 - $316.00
Chemical Identifiers
CAS | 20595-45-3 |
---|---|
Molecular Formula | C9H6Cl2O2 |
Molecular Weight (g/mol) | 217.045 |
MDL Number | MFCD00004373 |
InChI Key | MEBWABJHRAYGFW-DUXPYHPUSA-N |
Synonym | 2,4-dichlorocinnamic acid, trans-2,4-dichlorocinnamic acid, e-3-2,4-dichlorophenyl acrylic acid, cinnamic acid, 2,4-dichloro, 3-2,4-dichlorophenyl-2-propenoic acid, unii-c09kk5d2t2, 2-propenoic acid, 3-2,4-dichlorophenyl-, 2e, 2e-3-2,4-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,4-dichlorophenyl, 2e-3-2,4-dichlorophenyl prop-2-enoic acid |
PubChem CID | 688026 |
IUPAC Name | (E)-3-(2,4-dichlorophenyl)prop-2-enoic acid |
SMILES | C1=CC(=C(C=C1Cl)Cl)C=CC(=O)O |
Chemical Identifiers
20595-45-3 | |
217.045 | |
MEBWABJHRAYGFW-DUXPYHPUSA-N | |
688026 | |
C1=CC(=C(C=C1Cl)Cl)C=CC(=O)O |
C9H6Cl2O2 | |
MFCD00004373 | |
2,4-dichlorocinnamic acid, trans-2,4-dichlorocinnamic acid, e-3-2,4-dichlorophenyl acrylic acid, cinnamic acid, 2,4-dichloro, 3-2,4-dichlorophenyl-2-propenoic acid, unii-c09kk5d2t2, 2-propenoic acid, 3-2,4-dichlorophenyl-, 2e, 2e-3-2,4-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,4-dichlorophenyl, 2e-3-2,4-dichlorophenyl prop-2-enoic acid | |
(E)-3-(2,4-dichlorophenyl)prop-2-enoic acid |
Specifications
20595-45-3 | |
White | |
MFCD00004373 | |
2,4-dichlorocinnamic acid, trans-2,4-dichlorocinnamic acid, e-3-2,4-dichlorophenyl acrylic acid, cinnamic acid, 2,4-dichloro, 3-2,4-dichlorophenyl-2-propenoic acid, unii-c09kk5d2t2, 2-propenoic acid, 3-2,4-dichlorophenyl-, 2e, 2e-3-2,4-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,4-dichlorophenyl, 2e-3-2,4-dichlorophenyl prop-2-enoic acid | |
C1=CC(=C(C=C1Cl)Cl)C=CC(=O)O | |
217.045 | |
217.05 | |
Crystalline Powder |
236°C | |
C9H6Cl2O2 | |
5 g | |
MEBWABJHRAYGFW-DUXPYHPUSA-N | |
(E)-3-(2,4-dichlorophenyl)prop-2-enoic acid | |
688026 | |
≥98.0% (GC,T) | |
trans-2,4-Dichlorocinnamic Acid |
Safety and Handling
RTECSNumber : GD8575000
TSCA : No