missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-2-Aminocyclohexanol hydrochloride, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2232414

 View more versions of this product

Catalog No. AAB2232414

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

trans-2-Aminocyclohexanol hydrochloride
172°C to 175°C
MFCD00001490,MFCD09259962,MFCD09259962,MFCD09259963,MFCD11618002,MFCD11618003,MFCD09259963
Hygroscopic
SEBSRZIHRZRNAH-KGZKBUQUSA-N
(1R,2R)-2-aminocyclohexan-1-ol hydrochloridyl
12228412
≥98%
5456-63-3
C6H13ClNO
25 g
trans-2-aminocyclohexanol hydrochloride, 1r,2r-2-aminocyclohexanol hydrochloride, 1r, 2r-2-aminocyclohexanol hydrochloride, 1r,2r-2-aminocyclohexanol hcl, 1r,2r-trans-2-aminocyclohexanol hydrochloride, trans-2-hydroxycyclohexylamine hydrochloride, 1r,2r-2-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 2-amino-, hydrochloride, 1r,2r-rel, 1r,2r-2-aminocyclohexanolhydrochloride, 1r,2r-rel-2-aminocyclohexanol hydrochloride
[Cl].N[C@@H]1CCCC[C@H]1O
150.63
151.64

Chemical Identifiers

5456-63-3
150.63
SEBSRZIHRZRNAH-KGZKBUQUSA-N
(1R,2R)-2-aminocyclohexan-1-ol hydrochloridyl
C6H13ClNO
MFCD00001490,MFCD09259962,MFCD09259962,MFCD09259963,MFCD11618002,MFCD11618003,MFCD09259963
12228412
[Cl].N[C@@H]1CCCC[C@H]1O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only