missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hepten-1-ol 96.0+%, TCI America™

$315.03 - $315.03

Chemical Identifiers

CAS 33467-76-4
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00014056
InChI Key ASFYPVGAALGVNR-AATRIKPKSA-N
Synonym trans-2-hepten-1-ol, e-2-hepten-1-ol, e-hept-2-en-1-ol, 2-hepten-1-ol, e, 2-hepten-1-ol, 2e, 2-heptenol, unii-q989yfo10i, 2-hepten-1-ol, hept-2-en-1-ol, 2e-hept-2-en-1-ol
PubChem CID 5318017
IUPAC Name (E)-hept-2-en-1-ol
SMILES CCCCC=CCO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H06825ML
View Documents
TCI America
H06825ML
5 mL N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

33467-76-4
114.188
ASFYPVGAALGVNR-AATRIKPKSA-N
5318017
CCCCC=CCO
C7H14O
MFCD00014056
trans-2-hepten-1-ol, e-2-hepten-1-ol, e-hept-2-en-1-ol, 2-hepten-1-ol, e, 2-hepten-1-ol, 2e, 2-heptenol, unii-q989yfo10i, 2-hepten-1-ol, hept-2-en-1-ol, 2e-hept-2-en-1-ol
(E)-hept-2-en-1-ol

Specifications

33467-76-4
75°C
MFCD00014056
1987
ASFYPVGAALGVNR-AATRIKPKSA-N
(E)-hept-2-en-1-ol
5318017
≥96.0% (GC)
trans-2-Hepten-1-ol
Colorless
C7H14O
5 mL
trans-2-hepten-1-ol, e-2-hepten-1-ol, e-hept-2-en-1-ol, 2-hepten-1-ol, e, 2-hepten-1-ol, 2e, 2-heptenol, unii-q989yfo10i, 2-hepten-1-ol, hept-2-en-1-ol, 2e-hept-2-en-1-ol
CCCCC=CCO
114.188
114.19
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2393

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'