Learn More
trans-2-Hexenal, 99%, Thermo Scientific Chemicals
$80.86 - $80.86
Chemical Identifiers
CAS | 6728-26-3 |
---|---|
Molecular Formula | C6H10O |
Molecular Weight (g/mol) | 98.14 |
MDL Number | MFCD00007008 |
InChI Key | MBDOYVRWFFCFHM-SNAWJCMRSA-N |
Synonym | trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e |
PubChem CID | 5281168 |
ChEBI | CHEBI:28913 |
IUPAC Name | (E)-hex-2-enal |
SMILES | CCCC=CC=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC158130250
|
Thermo Scientific Chemicals
158130250 |
25 mL | Glass bottle |
Each for $80.86
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6728-26-3 | |
98.14 | |
MBDOYVRWFFCFHM-SNAWJCMRSA-N | |
5281168 | |
(E)-hex-2-enal |
C6H10O | |
MFCD00007008 | |
trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e | |
CHEBI:28913 | |
CCCC=CC=O |
Specifications
6728-26-3 | |
Colorless to Yellow | |
146.0°C to 149.0°C | |
Authentic | |
Glass bottle | |
1.4440 to 1.4500 | |
MFCD00007008 | |
01, I, 382 | |
trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e | |
MBDOYVRWFFCFHM-SNAWJCMRSA-N | |
(E)-hex-2-enal | |
5281168 | |
98.14 | |
Liquid |
5mg KOH/g max. | |
0.8460g/mL | |
35°C | |
98.5% min. (GC) | |
C6H10O | |
CH3CH2CH2CH=CHCHO | |
25 mL | |
0.846 | |
Solubility in water: insoluble. | |
CCCC=CC=O | |
98.14 | |
CHEBI:28913 | |
99% | |
trans-2-Hexenal, 99% |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
EINECSNumber : 229-778-1
RTECSNumber : MP5900000
TSCA : TSCA
RUO – Research Use Only