Learn More
trans-2-Hexenyl acetate, 98%, Thermo Scientific Chemicals
$70.01 - $237.01
Chemical Identifiers
CAS | 2497-18-9 |
---|---|
Molecular Formula | C8H14O2 |
Molecular Weight (g/mol) | 142.198 |
MDL Number | MFCD00009474 |
InChI Key | HRHOWZHRCRZVCU-AATRIKPKSA-N |
Synonym | z-hex-2-enyl acetate, unii-pt792bq57d, 2-hexen-1-ol, acetate, z, z-2-hexenyl acetate, fema no. 2564, 2-hexen-1-yl acetate, trans, 2-hexen-1-ol, acetate, 2e, 2-hexenyl acetate, trans, hexenyl acetate, 2z |
PubChem CID | 2733294 |
IUPAC Name | [(E)-hex-2-enyl] acetate |
SMILES | CCCC=CCOC(=O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1878618
|
Thermo Scientific Chemicals
A1878618 |
50 g |
N/A
|
|
|||||
AAA1878630
|
Thermo Scientific Chemicals
A1878630 |
250 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2497-18-9 | |
142.198 | |
HRHOWZHRCRZVCU-AATRIKPKSA-N | |
2733294 | |
CCCC=CCOC(=O)C |
C8H14O2 | |
MFCD00009474 | |
z-hex-2-enyl acetate, unii-pt792bq57d, 2-hexen-1-ol, acetate, z, z-2-hexenyl acetate, fema no. 2564, 2-hexen-1-yl acetate, trans, 2-hexen-1-ol, acetate, 2e, 2-hexenyl acetate, trans, hexenyl acetate, 2z | |
[(E)-hex-2-enyl] acetate |
Specifications
2497-18-9 | |
165°C to 166°C | |
C8H14O2 | |
MFCD00009474 | |
UN3272 | |
z-hex-2-enyl acetate, unii-pt792bq57d, 2-hexen-1-ol, acetate, z, z-2-hexenyl acetate, fema no. 2564, 2-hexen-1-yl acetate, trans, 2-hexen-1-ol, acetate, 2e, 2-hexenyl acetate, trans, hexenyl acetate, 2z | |
CCCC=CCOC(=O)C | |
142.198 | |
142.2 | |
trans-2-Hexenyl acetate |
0.9 | |
58°C (136°F) | |
1.427 | |
50 g | |
1721851 | |
HRHOWZHRCRZVCU-AATRIKPKSA-N | |
[(E)-hex-2-enyl] acetate | |
2733294 | |
98% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 219-680-7
RTECSNumber : MP8425000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only