missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-2-Octene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1260606

 View more versions of this product

Catalog No. AAL1260606


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

trans-2-Octene
-89°C to -86°C
122°C to 123°C
C8H16
MFCD00009532
UN3295
trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
CCCCC\C=C\C
112.22
112.22
13389-42-9
0.718
14°C (57°F)
1.413
5 g
1719497
ILPBINAXDRFYPL-HWKANZROSA-N
(E)-oct-2-ene
5364448
97%

Chemical Identifiers

13389-42-9
112.22
ILPBINAXDRFYPL-HWKANZROSA-N
5364448
CCCCC\C=C\C
C8H16
MFCD00009532
trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
(E)-oct-2-ene
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 236-463-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only