missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-2-Pentene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0942106

 View more versions of this product

Catalog No. AAL0942106

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

trans-2-Pentene
-136°C
36°C to 37°C
Petroleum-like
1.379
5 g
1718795
trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e
CCC=CC
70.135
70.14
646-04-8
0.649
−45°C (−49°F)
C5H10
MFCD00009384
UN3295
14,7123
QMMOXUPEWRXHJS-HWKANZROSA-N
(E)-pent-2-ene
5326161
99%

Chemical Identifiers

646-04-8
70.135
QMMOXUPEWRXHJS-HWKANZROSA-N
5326161
CCC=CC
C5H10
MFCD00009384
trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e
(E)-pent-2-ene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 211-461-4

RTECSNumber : SB2184500

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only