missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-3,4-Difluoro-4'-(4-n-propylcyclohexyl)biphenyl, 97%, Thermo Scientific™
$139.05 - $496.45
Chemical Identifiers
CAS | 85312-59-0 |
---|---|
Molecular Formula | C21H24F2 |
Molecular Weight (g/mol) | 314.42 |
MDL Number | MFCD09838999 |
InChI Key | VULXHDGYVHCLLN-UHFFFAOYSA-N |
Synonym | 4'-trans-4-propylcyclohexyl-3,4-difluorobiphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-propylcyclohexyl phenyl benzene, 3,4-difluoro-4'-1s,4r-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl biphenyl, 4-4-propylcyclohexyl-3',4'-difluorobiphenyl, trans-3,4-difluoro-4'-4-n-propylcyclohexyl biphenyl |
PubChem CID | 612306 |
IUPAC Name | 1,2-difluoro-4-[4-(4-propylcyclohexyl)phenyl]benzene |
SMILES | CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5535903
|
Thermo Scientific Chemicals
H5535903 |
1 g |
Each for $139.05
|
|
|||||
AAH5535906
|
Thermo Scientific Chemicals
H5535906 |
5 g |
Each for $496.45
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySparingly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
85312-59-0 | |
314.42 | |
VULXHDGYVHCLLN-UHFFFAOYSA-N | |
612306 | |
CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1 |
C21H24F2 | |
MFCD09838999 | |
4'-trans-4-propylcyclohexyl-3,4-difluorobiphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-propylcyclohexyl phenyl benzene, 3,4-difluoro-4'-1s,4r-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl biphenyl, 4-4-propylcyclohexyl-3',4'-difluorobiphenyl, trans-3,4-difluoro-4'-4-n-propylcyclohexyl biphenyl | |
1,2-difluoro-4-[4-(4-propylcyclohexyl)phenyl]benzene |
Specifications
85312-59-0 | |
C21H24F2 | |
1 g | |
Sparingly soluble in water. | |
CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1 | |
314.42 | |
314.42 | |
trans-3,4-Difluoro-4'-(4-n-propylcyclohexyl)biphenyl |
66°C to 68°C | |
MFCD09838999 | |
4'-trans-4-propylcyclohexyl-3,4-difluorobiphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-propylcyclohexyl phenyl benzene, 3,4-difluoro-4'-1s,4r-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl biphenyl, 4-4-propylcyclohexyl-3',4'-difluorobiphenyl, trans-3,4-difluoro-4'-4-n-propylcyclohexyl biphenyl | |
VULXHDGYVHCLLN-UHFFFAOYSA-N | |
1,2-difluoro-4-[4-(4-propylcyclohexyl)phenyl]benzene | |
612306 | |
97% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures