Learn More
trans-3-Heptene, 97%, Thermo Scientific Chemicals
$87.52 - $1222.12
Chemical Identifiers
CAS | 14686-14-7 |
---|---|
Molecular Formula | C7H14 |
Molecular Weight (g/mol) | 98.19 |
MDL Number | MFCD00009475 |
InChI Key | WZHKDGJSXCTSCK-FNORWQNLSA-N |
Synonym | trans-3-heptene, e-hept-3-ene, e-3-heptene, 3-heptene, e, 3-heptene, 3e, unii-o4br0a824i, hept-3-ene, cis-3-heptene, 3-heptene c,t, z-hept-3-ene |
PubChem CID | 5357259 |
SMILES | CCC\C=C\CC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5348203
|
Thermo Scientific Chemicals
H5348203 |
1 g |
Each for $87.52
|
|
|||||
AAH5348206
|
Thermo Scientific Chemicals
H5348206 |
5 g |
Each for $310.25
|
|
|||||
AAH5348214
|
Thermo Scientific Chemicals
H5348214 |
25 g |
Each for $1,222.12
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14686-14-7 | |
98.19 | |
WZHKDGJSXCTSCK-FNORWQNLSA-N | |
5357259 |
C7H14 | |
MFCD00009475 | |
trans-3-heptene, e-hept-3-ene, e-3-heptene, 3-heptene, e, 3-heptene, 3e, unii-o4br0a824i, hept-3-ene, cis-3-heptene, 3-heptene c,t, z-hept-3-ene | |
CCC\C=C\CC |
Specifications
14686-14-7 | |
0.698 | |
−1°C (30°F) | |
1.403 | |
1 g | |
trans-3-heptene, e-hept-3-ene, e-3-heptene, 3-heptene, e, 3-heptene, 3e, unii-o4br0a824i, hept-3-ene, cis-3-heptene, 3-heptene c,t, z-hept-3-ene | |
CCC\C=C\CC | |
98.19 | |
98.19 | |
trans-3-Heptene |
-137°C | |
94°C | |
C7H14 | |
MFCD00009475 | |
UN3295 | |
WZHKDGJSXCTSCK-FNORWQNLSA-N | |
(E)-hept-3-ene | |
5357259 | |
97% |
Safety and Handling
GHS H Statement
H225-H304-H315-H319-H335
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 238-728-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only