missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3438422

 View more versions of this product

Catalog No. AAH3438422


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid
49708-81-8
C13H15ClO2
4-4-chlorophenyl cyclohexanecarboxylic acid, trans-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4-chlorophenyl cyclohexane-1-carboxylic acid, e-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4'-chlorophenyl cyclohexancarboxylic acid, cis-4-4-chlorophenyl cyclohexanecarboxylic acid, atovaquone related compound 1, cyclohexanecarboxylic acid, 4-4-chlorophenyl-, trans, 1r,4r-4-4-chlorophenyl cyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 4-4-chlorophenyl
C1CC(CCC1C2=CC=C(C=C2)Cl)C(=O)O
238.711
238.71
249°C to 254°C
100 g
MFCD06797626
NXXDIEYTMQYWJU-UHFFFAOYSA-N
4-(4-chlorophenyl)cyclohexane-1-carboxylic acid
2735792
98%

Chemical Identifiers

49708-81-8
238.711
NXXDIEYTMQYWJU-UHFFFAOYSA-N
2735792
C1CC(CCC1C2=CC=C(C=C2)Cl)C(=O)O
C13H15ClO2
MFCD06797626
4-4-chlorophenyl cyclohexanecarboxylic acid, trans-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4-chlorophenyl cyclohexane-1-carboxylic acid, e-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4'-chlorophenyl cyclohexancarboxylic acid, cis-4-4-chlorophenyl cyclohexanecarboxylic acid, atovaquone related compound 1, cyclohexanecarboxylic acid, 4-4-chlorophenyl-, trans, 1r,4r-4-4-chlorophenyl cyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 4-4-chlorophenyl
4-(4-chlorophenyl)cyclohexane-1-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only