Learn More
trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5256606
Description
trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride acts as a pharmaceutical intermediate. It is used to prepare N-Boc-protected proline methyl ester of calix[4]arene, which is efficiently catalyzes the enantioselective aldol reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-4-Amino-N-Boc-L-proline methyl ester hydrochloride acts as a pharmaceutical intermediate. It is used to prepare N-Boc-protected proline methyl ester of calix[4]arene, which is efficiently catalyzes the enantioselective aldol reaction.
Notes
Store in a cool place. Moisture sensitive. Incompatible with oxidizing agents and acids.
Specifications
trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride | |
C11H21ClN2O4 | |
5 g | |
KFYCQKLSGMAVQH-WLYNEOFISA-N | |
1-O-tert-butyl 2-O-methyl (2S,4R)-4-aminopyrrolidine-1,2-dicarboxylate;hydrochloride | |
17750455 | |
97% |
334999-32-5 | |
MFCD03787929 | |
2s,4r-1-tert-butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, n-boc-trans-4-amino-l-proline methyl ester hydrochloride, 2s,4r-1-boc-4-amino-l-proline methyl ester hcl, 2s,4r-4-amino-1-boc-pyrrolidine-2-carboxylic acid methyl ester-hcl, 1-tert-butyl 2-methyl 2s,4r-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, n-boc-trans-4-amino-l-proline methyl ester hydrochloride salt, methyl trans-3-amino-1-boc-5-pyrrolidinecarboxylate hcl, boc-trans-4-amino-l-proline methyl ester hydrochloride salt, 2s,4r-1-boc-4-amino-l-proline methyl ester hydrochloride, n-boc-trans-4-amino-l-proline methyl ester, hydrochloride salt | |
[H+].[Cl-].COC(=O)[C@@H]1C[C@@H](N)CN1C(=O)OC(C)(C)C | |
280.75 | |
280.75 |
Chemical Identifiers
334999-32-5 | |
280.75 | |
KFYCQKLSGMAVQH-WLYNEOFISA-N | |
17750455 |
C11H21ClN2O4 | |
MFCD03787929 | |
2s,4r-1-tert-butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, n-boc-trans-4-amino-l-proline methyl ester hydrochloride, 2s,4r-1-boc-4-amino-l-proline methyl ester hcl, 2s,4r-4-amino-1-boc-pyrrolidine-2-carboxylic acid methyl ester-hcl, 1-tert-butyl 2-methyl 2s,4r-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, n-boc-trans-4-amino-l-proline methyl ester hydrochloride salt, methyl trans-3-amino-1-boc-5-pyrrolidinecarboxylate hcl, boc-trans-4-amino-l-proline methyl ester hydrochloride salt, 2s,4r-1-boc-4-amino-l-proline methyl ester hydrochloride, n-boc-trans-4-amino-l-proline methyl ester, hydrochloride salt | |
[H+].[Cl-].COC(=O)[C@@H]1C[C@@H](N)CN1C(=O)OC(C)(C)C |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only