missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-4-Methyl-beta-nitrostyrene, 98%, Thermo Scientific Chemicals

$135.71 - $135.71

Chemical Identifiers

CAS 5153-68-4
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00024828
InChI Key JSPNBERPFLONRX-VOTSOKGWSA-N
Synonym 1-methyl-4-2-nitrovinyl benzene, trans-4-methyl-beta-nitrostyrene, e-1-methyl-4-2-nitrovinyl benzene, 4-methyl-b-nitrostyrene, 4-methyl-beta-nitrostyrene, 1-methyl-4-2-nitroethenyl benzene, 1-methyl-4-e-2-nitroethenyl benzene, 4-methyl-.beta.-nitrostyrene, 4-methyl-.omega.-nitrostyrene, e-beta-nitro-4-methylstyrene
PubChem CID 674135
IUPAC Name 1-methyl-4-[(E)-2-nitroethenyl]benzene
SMILES CC1=CC=C(C=C1)C=C[N+](=O)[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1472106
View Documents
Thermo Scientific Chemicals
A1472106
5 g
Each for $135.71
 
Description

Description

trans-4-Methyl-β-nitrostyrene may be used as a reagent in the synthesis of N-benzylpyrrolomorphinans. It is also used as an organic intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Methyl-β-nitrostyrene may be used as a reagent in the synthesis of N-benzylpyrrolomorphinans. It is also used as an organic intermediate for pharmaceutical.

Solubility
Sparingly Soluble in water (0.070 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

5153-68-4
163.176
JSPNBERPFLONRX-VOTSOKGWSA-N
674135
CC1=CC=C(C=C1)C=C[N+](=O)[O-]
C9H9NO2
MFCD00024828
1-methyl-4-2-nitrovinyl benzene, trans-4-methyl-beta-nitrostyrene, e-1-methyl-4-2-nitrovinyl benzene, 4-methyl-b-nitrostyrene, 4-methyl-beta-nitrostyrene, 1-methyl-4-2-nitroethenyl benzene, 1-methyl-4-e-2-nitroethenyl benzene, 4-methyl-.beta.-nitrostyrene, 4-methyl-.omega.-nitrostyrene, e-beta-nitro-4-methylstyrene
1-methyl-4-[(E)-2-nitroethenyl]benzene

Specifications

5153-68-4
C9H9NO2
5 g
1-methyl-4-2-nitrovinyl benzene, trans-4-methyl-beta-nitrostyrene, e-1-methyl-4-2-nitrovinyl benzene, 4-methyl-b-nitrostyrene, 4-methyl-beta-nitrostyrene, 1-methyl-4-2-nitroethenyl benzene, 1-methyl-4-e-2-nitroethenyl benzene, 4-methyl-.beta.-nitrostyrene, 4-methyl-.omega.-nitrostyrene, e-beta-nitro-4-methylstyrene
JSPNBERPFLONRX-VOTSOKGWSA-N
1-methyl-4-[(E)-2-nitroethenyl]benzene
674135
98%
100°C to 104°C
MFCD00024828
1864209
Sparingly Soluble in water (0.070g/L) (25°C).
CC1=CC=C(C=C1)C=C[N+](=O)[O-]
163.176
163.18
trans-4-Methyl-beta-nitrostyrene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only