Learn More
trans-4-Methylcyclohexylamine, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 2523-55-9 |
---|---|
Molecular Formula | C7H15N |
Molecular Weight (g/mol) | 113.2 |
InChI Key | KSMVBYPXNKCPAJ-UHFFFAOYSA-N |
Synonym | 4-methylcyclohexylamine, trans-4-methylcyclohexylamine, 4-methylcyclohexanamine, trans-4-methylcyclohexanamine, cyclohexanamine, 4-methyl, cis-4-methylcyclohexylamine, p-methylcyclohexylamine, trans-4-methyl-cyclohexylamine, trans-4-methylcyclohexyl amine, 4-methyl-cyclohexylamine |
PubChem CID | 80604 |
IUPAC Name | 4-methylcyclohexan-1-amine |
SMILES | CC1CCC(CC1)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC455920050
|
Thermo Scientific Chemicals
455920050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2523-55-9 | |
113.2 | |
4-methylcyclohexylamine, trans-4-methylcyclohexylamine, 4-methylcyclohexanamine, trans-4-methylcyclohexanamine, cyclohexanamine, 4-methyl, cis-4-methylcyclohexylamine, p-methylcyclohexylamine, trans-4-methyl-cyclohexylamine, trans-4-methylcyclohexyl amine, 4-methyl-cyclohexylamine | |
4-methylcyclohexan-1-amine |
C7H15N | |
KSMVBYPXNKCPAJ-UHFFFAOYSA-N | |
80604 | |
CC1CCC(CC1)N |
Specifications
2523-55-9 | |
151.0°C to 154.0°C | |
98.5% min. (GC) | |
CH3C6H10NH2 | |
5g | |
4-methylcyclohexylamine, trans-4-methylcyclohexylamine, 4-methylcyclohexanamine, trans-4-methylcyclohexanamine, cyclohexanamine, 4-methyl, cis-4-methylcyclohexylamine, p-methylcyclohexylamine, trans-4-methyl-cyclohexylamine, trans-4-methylcyclohexyl amine, 4-methyl-cyclohexylamine | |
CC1CCC(CC1)N | |
113.2 | |
113.2 | |
trans-4-Methylcyclohexylamine |
0.8500g/mL | |
Authentic | |
C7H15N | |
1.4500 to 1.5480 | |
0.85 | |
KSMVBYPXNKCPAJ-UHFFFAOYSA-N | |
4-methylcyclohexan-1-amine | |
80604 | |
99% |
Safety and Handling
GHS H Statement Flammable liquid and vapour. Causes severe skin burns and eye damage.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
RUO â Research Use Only