Learn More
trans-4-Octene, 97%, Thermo Scientific Chemicals
$71.96 - $861.96
Chemical Identifiers
CAS | 14850-23-8 |
---|---|
Molecular Formula | C8H16 |
Molecular Weight (g/mol) | 112.216 |
MDL Number | MFCD00009476 |
InChI Key | IRUCBBFNLDIMIK-BQYQJAHWSA-N |
Synonym | trans-4-octene, e-4-octene, trans-n-4-octene, e-oct-4-ene, 4-octene, e, 4-octene, 4-octene, 4e, unii-fkz5737b1w, n-trans-4-octene, 4e-4-octene |
PubChem CID | 5357253 |
IUPAC Name | (E)-oct-4-ene |
SMILES | CCCC=CCCC |
Description
It is used in pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14850-23-8 | |
112.216 | |
IRUCBBFNLDIMIK-BQYQJAHWSA-N | |
5357253 | |
CCCC=CCCC |
C8H16 | |
MFCD00009476 | |
trans-4-octene, e-4-octene, trans-n-4-octene, e-oct-4-ene, 4-octene, e, 4-octene, 4-octene, 4e, unii-fkz5737b1w, n-trans-4-octene, 4e-4-octene | |
(E)-oct-4-ene |
Specifications
14850-23-8 | |
122°C to 123°C | |
Characteristic | |
1.412 | |
2 g | |
1719104 | |
Sparingly soluble in water(10.7,mg/L). | |
CCCC=CCCC | |
112.216 | |
112.22 | |
Liquid |
0.715 g/mL | |
21°C (70°F) | |
C8H16 | |
MFCD00009476 | |
UN3295 | |
trans-4-octene, e-4-octene, trans-n-4-octene, e-oct-4-ene, 4-octene, e, 4-octene, 4-octene, 4e, unii-fkz5737b1w, n-trans-4-octene, 4e-4-octene | |
IRUCBBFNLDIMIK-BQYQJAHWSA-N | |
(E)-oct-4-ene | |
5357253 | |
97% | |
trans-4-Octene |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 238-913-3
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only