missing translation for 'onlineSavingsMsg'
Learn More

trans-4-tert-Butylcyclohexanecarboxylic Acid 98.0+%, TCI America™

$160.30 - $596.49

Chemical Identifiers

CAS 943-29-3
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00466237
InChI Key QVQKEGYITJBHRQ-UHFFFAOYSA-N
Synonym trans-4-tert-butylcyclohexanecarboxylic acid, 4-tert-butylcyclohexanecarboxylic acid, cis-4-tert-butylcyclohexanecarboxylic acid, trans-4-tert-butyl cyclohexanecarboxylic acid, 4-tert-butyl cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, cis, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl, cis-4-tert-butylcyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, trans, qvqkegyitjbhrq-dtorhvgosa-n
PubChem CID 136759
IUPAC Name 4-tert-butylcyclohexane-1-carboxylic acid
SMILES CC(C)(C)C1CCC(CC1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30371G
View Documents
TCI America
B30371G
1 g
Each for $160.30
Only null left
 
B30375G
View Documents
TCI America
B30375G
5 g
Each for $596.49
Only null left
 
Description

Description

Specifications

Chemical Identifiers

943-29-3
184.279
QVQKEGYITJBHRQ-UHFFFAOYSA-N
136759
CC(C)(C)C1CCC(CC1)C(=O)O
C11H20O2
MFCD00466237
trans-4-tert-butylcyclohexanecarboxylic acid, 4-tert-butylcyclohexanecarboxylic acid, cis-4-tert-butylcyclohexanecarboxylic acid, trans-4-tert-butyl cyclohexanecarboxylic acid, 4-tert-butyl cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, cis, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl, cis-4-tert-butylcyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, trans, qvqkegyitjbhrq-dtorhvgosa-n
4-tert-butylcyclohexane-1-carboxylic acid

Specifications

943-29-3
White
MFCD00466237
trans-4-tert-butylcyclohexanecarboxylic acid, 4-tert-butylcyclohexanecarboxylic acid, cis-4-tert-butylcyclohexanecarboxylic acid, trans-4-tert-butyl cyclohexanecarboxylic acid, 4-tert-butyl cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, cis, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl, cis-4-tert-butylcyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, trans, qvqkegyitjbhrq-dtorhvgosa-n
CC(C)(C)C1CCC(CC1)C(=O)O
184.279
184.28
Crystalline Powder
176°C
C11H20O2
1 g
QVQKEGYITJBHRQ-UHFFFAOYSA-N
4-tert-butylcyclohexane-1-carboxylic acid
136759
≥98.0% (GC,T)
trans-4-tert-Butylcyclohexanecarboxylic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents