missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-Anethole, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 152530050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
trans-Anethole | |
4180-23-8 | |
Colorless to Yellow | |
234.0°C to 237.0°C | |
Authentic | |
Glass bottle | |
1.5590 to 1.5620 | |
MFCD00009284 | |
06, 566 | |
15, 636 | |
Solubility in water: practically insoluble. Other solubilities: miscible with ether, chloroform, soluble in benzene, ethyl acetate, acetone,, alcohol and carbon disulfide | |
CC=CC1=CC=C(C=C1)OC | |
148.2 | |
CHEBI:35616 | |
99% |
99% | |
23.0°C | |
0.9875g/mL | |
90°C | |
98.5% min. (GC) | |
C10H12O | |
CH3CH=CHC6H4OCH3 | |
5 mL | |
0.9875 | |
anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anethole | |
RUVINXPYWBROJD-ONEGZZNKSA-N | |
1-methoxy-4-[(E)-prop-1-enyl]benzene | |
637563 | |
148.2 | |
Liquid After Melting |
Chemical Identifiers
4180-23-8 | |
148.2 | |
RUVINXPYWBROJD-ONEGZZNKSA-N | |
637563 | |
1-methoxy-4-[(E)-prop-1-enyl]benzene |
C10H12O | |
MFCD00009284 | |
anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anethole | |
CHEBI:35616 | |
CC=CC1=CC=C(C=C1)OC |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 224-052-
RUO – Research Use Only