missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-Anethole, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 152535000

 View more versions of this product

Catalog No. AC152535000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

trans-Anethole
4180-23-8
Colorless to Yellow
234.0°C to 237.0°C
Authentic
Glass bottle
1.5590 to 1.5620
MFCD00009284
06, 566
15, 636
Solubility in water: practically insoluble. Other solubilities: miscible with ether, chloroform, soluble in benzene, ethyl acetate, acetone,, alcohol and carbon disulfide
CC=CC1=CC=C(C=C1)OC
148.2
CHEBI:35616
99%
99%
23.0°C
0.9875g/mL
90°C
98.5% min. (GC)
C10H12O
CH3CH=CHC6H4OCH3
500 mL
0.9875
anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anethole
RUVINXPYWBROJD-ONEGZZNKSA-N
1-methoxy-4-[(E)-prop-1-enyl]benzene
637563
148.2
Liquid After Melting

Chemical Identifiers

4180-23-8
148.2
RUVINXPYWBROJD-ONEGZZNKSA-N
637563
1-methoxy-4-[(E)-prop-1-enyl]benzene
C10H12O
MFCD00009284
anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anethole
CHEBI:35616
CC=CC1=CC=C(C=C1)OC
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 224-052-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only