missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 110350500

 View more versions of this product

Catalog No. AC110350500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

trans-Cinnamaldehyde
-7.5°C
1.0500g/mL
71°C
98.5% min. (GC)
C9H8O
C6H5CH=CHCHO
50 g
1.05
cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
KJPRLNWUNMBNBZ-QPJJXVBHSA-N
(E)-3-phenylprop-2-enal
637511
132.16
Liquid
14371-10-9
Yellow
250.0°C to 252.0°C
Authentic
Glass bottle
1.6210 to 1.6230
MFCD00007000
07,348
14,2297
Solubility in water: 1.1g/L (20°C) (kolbenmethode).
O=C\C=C\C1=CC=CC=C1
132.16
CHEBI:16731
99%

Chemical Identifiers

14371-10-9
132.16
KJPRLNWUNMBNBZ-QPJJXVBHSA-N
637511
O=C\C=C\C1=CC=CC=C1
C9H8O
MFCD00007000
cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
CHEBI:16731
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only