missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals

$56.37 - $170.45

Chemical Identifiers

CAS 140-10-3
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00004369
InChI Key WBYWAXJHAXSJNI-VOTSOKGWSA-N
Synonym cinnamic acid, trans-cinnamic acid, e-cinnamic acid, 3-phenylacrylic acid, trans-3-phenylacrylic acid, zimtsaeure, 2e-3-phenylprop-2-enoic acid, 3-phenylprop-2-enoic acid, 3-phenylpropenoic acid
PubChem CID 444539
ChEBI CHEBI:35697
IUPAC Name (E)-3-phenylprop-2-enoic acid
SMILES C1=CC=C(C=C1)C=CC(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158570050
SDS View Documents
Thermo Scientific Chemicals
158570050
5 g Glass bottle
Each for $56.37
 
AC158571000
SDS View Documents
Thermo Scientific Chemicals
158571000
100 g Plastic bottle
Each for $58.99
 
AC158575000
SDS View Documents
Thermo Scientific Chemicals
158575000
500 g Plastic bottle
Each for $170.45
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

140-10-3
148.161
WBYWAXJHAXSJNI-VOTSOKGWSA-N
444539
(E)-3-phenylprop-2-enoic acid
C9H8O2
MFCD00004369
cinnamic acid, trans-cinnamic acid, e-cinnamic acid, 3-phenylacrylic acid, trans-3-phenylacrylic acid, zimtsaeure, 2e-3-phenylprop-2-enoic acid, 3-phenylprop-2-enoic acid, 3-phenylpropenoic acid
CHEBI:35697
C1=CC=C(C=C1)C=CC(=O)O

Specifications

131.0°C to 136.0°C
300.0°C
98+%
C6H5CH=CHCO2H
09, 573
cinnamic acid, trans-cinnamic acid, e-cinnamic acid, 3-phenylacrylic acid, trans-3-phenylacrylic acid, zimtsaeure, 2e-3-phenylprop-2-enoic acid, 3-phenylprop-2-enoic acid, 3-phenylpropenoic acid
WBYWAXJHAXSJNI-VOTSOKGWSA-N
(E)-3-phenylprop-2-enoic acid
444539
148.16
1.2480g/mL
>100°C
Glass bottle
1.248
140-10-3
White
C9H8O2
MFCD00004369
14, 2299
Solubility in water: 0.4g/L in water (20°C). Other solubilities: soluble in glacial acetic acid, 1g/6mL alcohol - 1g/5mL methanol, 1g/12mL chloroform, soluble in benzene, ether, acetone, oils, and carbon disulfide
C1=CC=C(C=C1)C=CC(=O)O
148.161
CHEBI:35697
≥98%
5 g
Authentic
Crystalline Powder
trans-Cinnamic acid, 98+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 205-398-1

RTECSNumber : GD7850000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only