Learn More
trans-Crotonic acid, 98%, Thermo Scientific Chemicals
$38.41 - $391.95
Chemical Identifiers
CAS | 107-93-7 |
---|---|
Molecular Formula | C4H6O2 |
Molecular Weight (g/mol) | 86.09 |
MDL Number | MFCD00002701 |
InChI Key | LDHQCZJRKDOVOX-NSCUHMNNSA-N |
Synonym | crotonic acid, e-but-2-enoic acid, but-2-enoic acid, trans-2-butenoic acid, trans-crotonic acid, 3-methylacrylic acid, e-2-butenoic acid, 2-butenoic acid, 2e-but-2-enoic acid, e-crotonic acid |
PubChem CID | 637090 |
ChEBI | CHEBI:41131 |
IUPAC Name | (E)-but-2-enoic acid |
SMILES | CC=CC(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1576514
|
Thermo Scientific Chemicals
A1576514 |
25 g |
Each for $38.41
|
|
|||||
AAA1576536
|
Thermo Scientific Chemicals
A1576536 |
500 g |
Each for $101.63
|
|
|||||
AAA157650E
|
Thermo Scientific Chemicals
A157650E |
2500 g |
Each for $391.95
|
|
|||||
Description
trans-Crotonic acid is used in the copolymerization of crotonic acid hydrogel systems by using gamma-rays. The crosslinked copolymeric crotonic acid hydrogels release fertilizers and drugs that help to prevent environmental pollution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-Crotonic acid is used in the copolymerization of crotonic acid hydrogel systems by using gamma-rays. The crosslinked copolymeric crotonic acid hydrogels release fertilizers and drugs that help to prevent environmental pollution.
Solubility
Soluble in water.
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Chemical Identifiers
107-93-7 | |
86.09 | |
LDHQCZJRKDOVOX-NSCUHMNNSA-N | |
637090 | |
(E)-but-2-enoic acid |
C4H6O2 | |
MFCD00002701 | |
crotonic acid, e-but-2-enoic acid, but-2-enoic acid, trans-2-butenoic acid, trans-crotonic acid, 3-methylacrylic acid, e-2-butenoic acid, 2-butenoic acid, 2e-but-2-enoic acid, e-crotonic acid | |
CHEBI:41131 | |
CC=CC(=O)O |
Specifications
107-93-7 | |
1.027 | |
87°C (188°F) | |
MFCD00002701 | |
UN2823 | |
14,2597 | |
Soluble in water. | |
CC=CC(=O)O | |
86.09 | |
CHEBI:41131 | |
98% |
70°C to 74°C | |
128°C (100 mmHg) | |
C4H6O2 | |
25 g | |
1719943 | |
crotonic acid, e-but-2-enoic acid, but-2-enoic acid, trans-2-butenoic acid, trans-crotonic acid, 3-methylacrylic acid, e-2-butenoic acid, 2-butenoic acid, 2e-but-2-enoic acid, e-crotonic acid | |
LDHQCZJRKDOVOX-NSCUHMNNSA-N | |
(E)-but-2-enoic acid | |
637090 | |
86.09 | |
trans-Crotonic acid |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P280-P305+P351+P338-P310
H318
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CROTONIC ACID, SOLID
EINECSNumber : 203-533-9
RTECSNumber : GQ2800000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only