missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-N,N'-Diacetylcyclohexane-1,2-diamine 98.0+%, TCI America™
$190.13 - $518.21
Chemical Identifiers
| CAS | 70924-78-6 |
|---|---|
| Molecular Formula | C10H18N2O2 |
| Molecular Weight (g/mol) | 198.266 |
| InChI Key | MZYLAHHTXWRMOL-NXEZZACHSA-N |
| Synonym | trans-1,2-Bis(acetamido)cyclohexane |
| PubChem CID | 11805689 |
| IUPAC Name | N-[(1R,2R)-2-acetamidocyclohexyl]acetamide |
| SMILES | CC(=O)NC1CCCCC1NC(=O)C |
Chemical Identifiers
| 70924-78-6 | |
| 198.266 | |
| trans-1,2-Bis(acetamido)cyclohexane | |
| N-[(1R,2R)-2-acetamidocyclohexyl]acetamide |
| C10H18N2O2 | |
| MZYLAHHTXWRMOL-NXEZZACHSA-N | |
| 11805689 | |
| CC(=O)NC1CCCCC1NC(=O)C |
Specifications
| 70924-78-6 | |
| White-Yellow | |
| 5 g | |
| MZYLAHHTXWRMOL-NXEZZACHSA-N | |
| N-[(1R,2R)-2-acetamidocyclohexyl]acetamide | |
| 11805689 | |
| ≥98.0% (GC) | |
| trans-N,N′-Diacetylcyclohexane-1,2-diamine |
| 270°C | |
| C10H18N2O2 | |
| trans-1,2-Bis(acetamido)cyclohexane | |
| CC(=O)NC1CCCCC1NC(=O)C | |
| 198.266 | |
| 198.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : No