missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans,trans-1,5-Diphenyl-1,4-pentadien-3-one 99.0+%, TCI America™
Supplier: TCI America D0903250G
Specifications
trans,trans-1,5-Diphenyl-1,4-pentadien-3-one | |
113°C | |
MFCD00004790 | |
dibenzylideneacetone, 1,5-diphenylpenta-1,4-dien-3-one, trans,trans-dibenzylideneacetone, dibenzalacetone, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one, dibenzylidene acetone, distyryl ketone, 1,5-diphenyl-1,4-pentadien-3-one, trans,trans-dibenzalacetone, trans,trans-1,5-diphenyl-1,4-pentadien-3-one | |
O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1 | |
234.30 | |
234.30 | |
Crystalline Powder |
35225-79-7 | |
C17H14O | |
250 g | |
WMKGGPCROCCUDY-PHEQNACWSA-N | |
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one | |
640180 | |
≥99.0% (GC) |
Chemical Identifiers
35225-79-7 | |
234.30 | |
WMKGGPCROCCUDY-PHEQNACWSA-N | |
640180 | |
O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1 |
C17H14O | |
MFCD00004790 | |
dibenzylideneacetone, 1,5-diphenylpenta-1,4-dien-3-one, trans,trans-dibenzylideneacetone, dibenzalacetone, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one, dibenzylidene acetone, distyryl ketone, 1,5-diphenyl-1,4-pentadien-3-one, trans,trans-dibenzalacetone, trans,trans-1,5-diphenyl-1,4-pentadien-3-one | |
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one |
Safety and Handling
TSCA : Yes