missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans,trans-2,4-Hexadien-1-ol, 99%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 325570500

 View more versions of this product

Catalog No. AC325570500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

trans, trans-2, 4-Hexadien-1-ol
30.5°C to 31.5°C
0.8700g/mL
72°C
98.5% min. (GC)
C6H10O
CH3CH=CHCH=CHCH2OH
50 g
15,8848
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and oils
C\C=C\C=C\CO
98.15
641256
99%
17102-64-6
Colorless to Yellow
80.0°C (12.0 mmHg)
Authentic
Glass bottle
1.4990 to 1.5010
MFCD00002925
0.87
sorbic alcohol, 2,4-hexadien-1-ol, sorbyl alcohol, trans,trans-2,4-hexadien-1-ol, 2,4-hexadienol, hexacose, hexakose, sorbinic alcohol, 1-hydroxy-2,4-hexadiene, 2e,4e-hexa-2,4-dien-1-ol
MEIRRNXMZYDVDW-MQQKCMAXSA-N
(2E,4E)-hexa-2,4-dien-1-ol
0.1% alpha-tocopherol
98.14
Liquid After Melting

Chemical Identifiers

17102-64-6
98.15
MEIRRNXMZYDVDW-MQQKCMAXSA-N
641256
C\C=C\C=C\CO
C6H10O
MFCD00002925
sorbic alcohol, 2,4-hexadien-1-ol, sorbyl alcohol, trans,trans-2,4-hexadien-1-ol, 2,4-hexadienol, hexacose, hexakose, sorbinic alcohol, 1-hydroxy-2,4-hexadiene, 2e,4e-hexa-2,4-dien-1-ol
(2E,4E)-hexa-2,4-dien-1-ol
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 241-173-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only