missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-propylbicyclohexyl 98.0+%, TCI America™

Supplier:  TCI America E11571G

 View more versions of this product

Catalog No. E11571G



Description

Description

Specifications

Specifications

trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4′-propylbicyclohexyl
79°C
C23H34F2O
1 g
CFJCNWQXOICKIF-UHFFFAOYSA-N
1-ethoxy-2,3-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene
11417199
≥98.0% (GC)
123560-48-5
White
MFCD18433524
trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, trans,trans-4'-4-ethoxy-2,3-difluoro-phenyl-4-propyl-bicyclohexyl, trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-n-propylbicyclohexyl, 1r,1'r,4r,4's-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1-ethoxy-2,3-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-propylbicyclohexyl, trans-4-4-ethoxy-2,3-difluorophenyl-trans-4'-propylbicyclohexane, benzene, 1-ethoxy-2,3-difluoro-4-4'-propyl 1,1'-bicyclohexyl-4-yl, trans-4-4-ethoxy-2,3-difluoro-phenyl-4-propyl-bicycloh, 1r,1's,4r,4'r-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane
CCCC1CCC(CC1)C2CCC(CC2)C3=C(C(=C(C=C3)OCC)F)F
364.521
364.52
Crystalline Powder

Chemical Identifiers

123560-48-5
364.521
CFJCNWQXOICKIF-UHFFFAOYSA-N
11417199
CCCC1CCC(CC1)C2CCC(CC2)C3=C(C(=C(C=C3)OCC)F)F
C23H34F2O
MFCD18433524
trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, trans,trans-4'-4-ethoxy-2,3-difluoro-phenyl-4-propyl-bicyclohexyl, trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-n-propylbicyclohexyl, 1r,1'r,4r,4's-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1-ethoxy-2,3-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-propylbicyclohexyl, trans-4-4-ethoxy-2,3-difluorophenyl-trans-4'-propylbicyclohexane, benzene, 1-ethoxy-2,3-difluoro-4-4'-propyl 1,1'-bicyclohexyl-4-yl, trans-4-4-ethoxy-2,3-difluoro-phenyl-4-propyl-bicycloh, 1r,1's,4r,4'r-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane
1-ethoxy-2,3-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions