missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-propylbicyclohexyl 98.0+%, TCI America™
Supplier: TCI America E11575G
Specifications
trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4′-propylbicyclohexyl | |
79°C | |
C23H34F2O | |
5 g | |
CFJCNWQXOICKIF-UHFFFAOYSA-N | |
1-ethoxy-2,3-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene | |
11417199 | |
≥98.0% (GC) |
123560-48-5 | |
White | |
MFCD18433524 | |
trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, trans,trans-4'-4-ethoxy-2,3-difluoro-phenyl-4-propyl-bicyclohexyl, trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-n-propylbicyclohexyl, 1r,1'r,4r,4's-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1-ethoxy-2,3-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-propylbicyclohexyl, trans-4-4-ethoxy-2,3-difluorophenyl-trans-4'-propylbicyclohexane, benzene, 1-ethoxy-2,3-difluoro-4-4'-propyl 1,1'-bicyclohexyl-4-yl, trans-4-4-ethoxy-2,3-difluoro-phenyl-4-propyl-bicycloh, 1r,1's,4r,4'r-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane | |
CCCC1CCC(CC1)C2CCC(CC2)C3=C(C(=C(C=C3)OCC)F)F | |
364.521 | |
364.52 | |
Crystalline Powder |
Chemical Identifiers
123560-48-5 | |
364.521 | |
CFJCNWQXOICKIF-UHFFFAOYSA-N | |
11417199 | |
CCCC1CCC(CC1)C2CCC(CC2)C3=C(C(=C(C=C3)OCC)F)F |
C23H34F2O | |
MFCD18433524 | |
trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, trans,trans-4'-4-ethoxy-2,3-difluoro-phenyl-4-propyl-bicyclohexyl, trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-n-propylbicyclohexyl, 1r,1'r,4r,4's-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1-ethoxy-2,3-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-propylbicyclohexyl, trans-4-4-ethoxy-2,3-difluorophenyl-trans-4'-propylbicyclohexane, benzene, 1-ethoxy-2,3-difluoro-4-4'-propyl 1,1'-bicyclohexyl-4-yl, trans-4-4-ethoxy-2,3-difluoro-phenyl-4-propyl-bicycloh, 1r,1's,4r,4'r-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane | |
1-ethoxy-2,3-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene |
Safety and Handling
TSCA : No