missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-4'-Propyl-4-(p-tolyl)bicyclohexyl 98.0+%, TCI America™
Supplier: TCI America P24041G
Specifications
| trans,trans-4′-Propyl-4-(p-tolyl)bicyclohexyl | |
| 110°C | |
| C22H34 | |
| 1 g | |
| SDESCXGEQILYTQ-UHFFFAOYSA-N | |
| 4-(4-methylphenyl)-4'-propyl-1,1'-bi(cyclohexane) | |
| 576378 | |
| ≥98.0% (GC) |
| 84656-75-7 | |
| White | |
| MFCD13182311 | |
| 1-methyl-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl benzene, trans,trans-4'-p-tolyl-4-propyl-bicyclohexyl, benzene, 1-methyl-4-4'-propyl 1,1'-bicyclohexyl-4-yl, trans,trans-4-4-methylphenyl-4'-n-propylbicyclohexyl, 4-trans-4 trans-4-propylcyclohexyl cyclohexyl toluene, trans,trans-4-p-tolyl-4-propyl-bicyclohexyl, trans,trans-4'-propyl-4-p-tolyl bicyclohexyl, 4-propyl-4'-p-tolyl-1,1'-bi cyclohexane, 1-methyl-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4'-propyl-4-4-methylphenyl bicyclohexyl | |
| CCCC1CCC(CC1)C1CCC(CC1)C1=CC=C(C)C=C1 | |
| 298.51 | |
| 298.51 | |
| Crystalline Powder |
Chemical Identifiers
| 84656-75-7 | |
| 298.51 | |
| SDESCXGEQILYTQ-UHFFFAOYSA-N | |
| 576378 | |
| CCCC1CCC(CC1)C1CCC(CC1)C1=CC=C(C)C=C1 |
| C22H34 | |
| MFCD13182311 | |
| 1-methyl-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl benzene, trans,trans-4'-p-tolyl-4-propyl-bicyclohexyl, benzene, 1-methyl-4-4'-propyl 1,1'-bicyclohexyl-4-yl, trans,trans-4-4-methylphenyl-4'-n-propylbicyclohexyl, 4-trans-4 trans-4-propylcyclohexyl cyclohexyl toluene, trans,trans-4-p-tolyl-4-propyl-bicyclohexyl, trans,trans-4'-propyl-4-p-tolyl bicyclohexyl, 4-propyl-4'-p-tolyl-1,1'-bi cyclohexane, 1-methyl-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4'-propyl-4-4-methylphenyl bicyclohexyl | |
| 4-(4-methylphenyl)-4'-propyl-1,1'-bi(cyclohexane) |
Safety and Handling
EINECSNumber : (4)-1985
TSCA : No