missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-Dibenzylideneacetone, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 35225-79-7 |
|---|---|
| Molecular Formula | C17H14O |
| Molecular Weight (g/mol) | 234.3 |
| MDL Number | MFCD00004790 |
| InChI Key | WMKGGPCROCCUDY-PHEQNACWSA-N |
| Synonym | dibenzylideneacetone, 1,5-diphenylpenta-1,4-dien-3-one, trans,trans-dibenzylideneacetone, dibenzalacetone, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one, dibenzylidene acetone, distyryl ketone, 1,5-diphenyl-1,4-pentadien-3-one, trans,trans-dibenzalacetone, trans,trans-1,5-diphenyl-1,4-pentadien-3-one |
| PubChem CID | 640180 |
| IUPAC Name | (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one |
| SMILES | C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC409000050
|
Thermo Scientific Chemicals
409000050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC409000250
|
Thermo Scientific Chemicals
409000250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC409001000
|
Thermo Scientific Chemicals
409001000 |
100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 35225-79-7 | |
| 234.3 | |
| WMKGGPCROCCUDY-PHEQNACWSA-N | |
| 640180 | |
| C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2 |
| C17H14O | |
| MFCD00004790 | |
| dibenzylideneacetone, 1,5-diphenylpenta-1,4-dien-3-one, trans,trans-dibenzylideneacetone, dibenzalacetone, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one, dibenzylidene acetone, distyryl ketone, 1,5-diphenyl-1,4-pentadien-3-one, trans,trans-dibenzalacetone, trans,trans-1,5-diphenyl-1,4-pentadien-3-one | |
| (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one |
Specifications
| 35225-79-7 | |
| 97.5% min | |
| C17H14O | |
| MFCD00004790 | |
| 07, IV, 1747 | |
| dibenzylideneacetone, 1,5-diphenylpenta-1,4-dien-3-one, trans,trans-dibenzylideneacetone, dibenzalacetone, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one, dibenzylidene acetone, distyryl ketone, 1,5-diphenyl-1,4-pentadien-3-one, trans,trans-dibenzalacetone, trans,trans-1,5-diphenyl-1,4-pentadien-3-one | |
| WMKGGPCROCCUDY-PHEQNACWSA-N | |
| (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one | |
| 640180 | |
| 98% |
| Authentic | |
| Glass bottle | |
| C6H5CH=CHC(O)CH=CHC6H5 | |
| 5g | |
| 15, 3010 | |
| Solubility in water: negliglible. Other solubilities: soluble in acetone and chloroform,slightly soluble in alcohol and ether | |
| C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2 | |
| 234.3 | |
| 234.3 | |
| trans, trans-Dibenzylideneacetone, 95% |
RUO â Research Use Only