BrandTech™ Transferpette™ -8/-12 electronic Multichannel Pipette - TRADE-UP PROMO
Combines comfortable design, balanced weight distribution, intuitive software and user-friendly documentation for your pipetting needs. PROMO - Save when you upgrade your old pipette to the Brand™ Transferpette™ pipette.
Supplier:  BrandTech™ 705476TU
 View more versions of this product
Catalog No. 14389132TU
Explore available promotions
Description
Description
  • Choice of 8 or 12 channel models
  • Clear, legible LCD screen
  • Intuitive interface with user-friendly desing allows easy operation
  • Soft touch pipetting key and adjustable finger rest provides added comfort
  • Rechargeable battery provides over 4000 pipetting cycles between charges
  • Multichannel V-rings on tip cones improves tip fit and reduces mounting and ejection force
  • Easily maintained
Operating modes
  • Pipetting (PIP Mode): previously set volume is aspirated into pipette tip and then discharged
  • Mixing of samples (PIPmix Mode): program for mixing liquids
  • Reverse Pipetting (revPIP Mode): program designed for greater accuracy
  • Pipetting for electrophoresis (GEL Mode): program for loading of electrophoresis gels
  • Dispensing (DISP Mode): single aspirated volume is dispensed in selected number of smaller, equal aliquots
Specifications
Specifications
±1.8μL
2 to 200μL tip, 5 to 300μL tip, 5 to 100μL tip, 5 to 200μL tip
12
Step Based
15 to 300 μL
Ergonomic
0.2μL
1 Year
Rechargeable
Transferpette™ -8/-12 Electronic
Transferpette™
Electronic Pipette
LCD
Battery, power supply unit, 120VAC adapter with charging cable, one pipette tip sample, operating manual, pipette stand, tip-box SL filled with appropriate tip, two tip refill inserts, reagent reservoir and five operating modes
Yes