missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Erlab™ Transparent Back Panel For Captair 321 Fume Hood
Clear back panel made of synthetic glass offering 360° visibility for 321 fume hood models.
Supplier: Erlab™ 321EDU
Description
- For use with Captair 321 Ductless Filtering Fume Hood
- Includes warranty
- Useful for teaching and student observation
Specifications
Captair Ductless Filtering Fume Hoods or CaptairFlow Laminar Flow Hoods | |
5.4 Kg | |
Back Panel | |
Teaching Observation or Locating in Front of Window | |
Rounded Edge |
12 lb. | |
Educational Back Panel and Thumb Screws | |
Acrylic | |
Clear | |
One Year |