Fisherbrand™ Transport Boxes
For transporting biological and clinical samples, as well as instruments and products that you want to keep clean and dry under testing environmental conditions. Fisherbrand™ Transport box is made of polycarbonate and available in three colors for user/owner identification.
$232.81 - $232.81
Products 1
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
12-006-916
View Documents
Fisherbrand™
120374
Clear, Blue
Each for $232.81
Case of 12 Each for $2,793.72
Description
Description
  • Polycarbonate construction and available in three colors for user/owner identification
  • Silicone water-tight seal and three lid clasps provide secure closure
  • Carry handle folds neatly into body space allowing the units to be stacked
  • Lid opens a full 180°, allowing total access to contents and making it easier to clean
  • Accommodates 13 and 16mm tubes in a 72-place onerack™, or similar sized/brand of rack
  • Separators provided to create own compartment sizes, or use empty to transport larger products and equipment
  • Autoclavable
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'