Innovatek Transwab™ Liquid Culture Swabs
Transwab™ Liquid Culture Swabs provide superior recovery of pathogens in a variety of applications
Supplier:  Innovatek 4080MW169P
 View more versions of this product
Catalog No. 361045657
Explore available promotions
Description
Description
  • Swab and labeled tube in peel pouch
  • Semi-solid or liquid transport media for superior recovery of pathogens
  • For use in medical, clinical, food, forensic, environmental, and industrial applications
  • Choose tip material and size, shaft material, liquid or gel transport media to suit every need
Specifications
Specifications
Amies Clear
Multiple Swab Tests, Multiple Sampling Sites
Plastic
Dual Applicator
125 Swabs/Box
Rayon