Learn More
Tri-n-butyltin chloride, 95%, tech., Thermo Scientific Chemicals
$99.05 - $402.02
Chemical Identifiers
CAS | 1461-22-9 |
---|---|
Molecular Formula | C12H27ClSn |
Molecular Weight (g/mol) | 325.48 |
MDL Number | MFCD00000521 |
InChI Key | GCTFWCDSFPMHHS-UHFFFAOYSA-M |
Synonym | tributyltin chloride, chlorotributyltin, tri-n-butyltin chloride, tributylchlorotin, stannane, tributylchloro, chlorotri-n-butyltin, tri-n-butylchlorotin, tributyl chloro stannane, chlorotributylstannane, tributylstannyl chloride |
PubChem CID | 15096 |
ChEBI | CHEBI:79734 |
IUPAC Name | tributyl(chloro)stannane |
SMILES | CCCC[Sn](CCCC)(CCCC)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC139351000
|
Thermo Scientific Chemicals
139351000 |
100 g | Glass bottle |
Each for $99.05
|
|
||||
AC139355000
|
Thermo Scientific Chemicals
139355000 |
500 g | Glass bottle |
Each for $402.02
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1461-22-9 | |
325.48 | |
GCTFWCDSFPMHHS-UHFFFAOYSA-M | |
15096 | |
tributyl(chloro)stannane |
C12H27ClSn | |
MFCD00000521 | |
tributyltin chloride, chlorotributyltin, tri-n-butyltin chloride, tributylchlorotin, stannane, tributylchloro, chlorotri-n-butyltin, tri-n-butylchlorotin, tributyl chloro stannane, chlorotributylstannane, tributylstannyl chloride | |
CHEBI:79734 | |
CCCC[Sn](CCCC)(CCCC)Cl |
Specifications
1461-22-9 | |
Colorless to Yellow | |
171.0°C to 173.0°C (25.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4880 to 1.4940 | |
MFCD00000521 | |
1.2 | |
tributyltin chloride, chlorotributyltin, tri-n-butyltin chloride, tributylchlorotin, stannane, tributylchloro, chlorotri-n-butyltin, tri-n-butylchlorotin, tributyl chloro stannane, chlorotributylstannane, tributylstannyl chloride | |
GCTFWCDSFPMHHS-UHFFFAOYSA-M | |
tributyl(chloro)stannane | |
15096 | |
CHEBI:79734 | |
95% | |
Liquid |
-9.0°C | |
1.2000g/mL | |
>112°C | |
94% min. (Argentometry) | |
C12H27ClSn | |
[CH3(CH2)3]3SnCl | |
100 g | |
15, 9782 | |
Solubility in water: 0.07g/L (20°C). Other solubilities: soluble in most common orgnaic solvents,including,benzene and toluene | |
CCCC[Sn](CCCC)(CCCC)Cl | |
325.48 | |
7 mPa.s (20°C) | |
325.48 | |
Technical | |
Tri-n-butyltin chloride, tech. 90% |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
Ca
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clo
GHS Signal Word: Danger
EINECSNumber : 215-958-7
RTECSNumber : WH6820000
TSCA : TSCA
RUO – Research Use Only